accessibility menu, dialog, popup

UserName

CAS RN 55289-06-0

IUPAC Name: 3-methoxy-2-methylbenzoic acid
Synonyms: 3-methoxy-2-methylbenzoic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: JPCISVSOTKMFPG-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C(O)=O)=C1C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)