accessibility menu, dialog, popup

UserName

CAS RN 55268-74-1

IUPAC Name: 2-cyclohexanecarbonyl-1H,2H,3H,4H,6H,7H,11bH-pyrazino[2,1-a]isoquinolin-4-one
Synonyms: praziquantel (R)-isomer praziquantel (+-)-praziquantel praziquantel, (+-)- traziquantel praziquantel, (+-)-isomer bayer brand of praziquantel praziquantel, (r)- praziquantel, (s)- praziquantel, (S)-isomer
Molecular Weight (g/mol): 312.41
Molecular Formula: C19H24N2O2
InChi Key: FSVJFNAIGNNGKK-UHFFFAOYNA-N
SMILES: O=C(C1CCCCC1)N1CC2N(CCC3=CC=CC=C23)C(=O)C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)