accessibility menu, dialog, popup

UserName

OPOO

CAS RN 5518-52-5

IUPAC Name: tris(furan-2-yl)phosphane
Synonyms: tris(furan-2-yl)phosphane
Molecular Weight (g/mol): 232.18
Molecular Formula: C12H9O3P
InChi Key: DLQYXUGCCKQSRJ-UHFFFAOYSA-N
SMILES: O1C=CC=C1P(C1=CC=CO1)C1=CC=CO1
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (4)
  • (3)
Quantity 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (3)
  • (1)
  • (3)