missing translation for 'onlineSavingsMsg'
Learn More

Tri-(2-furyl)phosphine, 98%

Catalog No. AC294800050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294800050 5 g Glass bottle
AC294800010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC294800050 Supplier Thermo Scientific Chemicals Supplier No. 294800050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5518-52-5 | C12H9O3P | 232.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5518-52-5
Molecular Formula C12H9O3P
Molecular Weight (g/mol) 232.18
MDL Number MFCD00151857
InChI Key DLQYXUGCCKQSRJ-UHFFFAOYSA-N
Synonym tri 2-furyl phosphine, tri furan-2-yl phosphine, tri-2-furyl phosphine, tris furan-2-yl phosphane, tri-2-furylphosphine, tris 2-furyl phosphine, tri furan-2-yl phosphane, trifurylphosphine, tri fur-2-yl phosphane, tris 2-furanyl phosphine
PubChem CID 521585
IUPAC Name tris(furan-2-yl)phosphane
SMILES O1C=CC=C1P(C1=CC=CO1)C1=CC=CO1

Specifications

Melting Point 61.0°C to 64.0°C
Boiling Point 136.0°C (4.0 mmHg)
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 232.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tri-(2-furyl)phosphine, 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H StatementCauses skin irritation.Causes serious eye irritation.May cause respiratory irritation.
Recommended Storage Refrigerator +4°C

RUO – Research Use Only