missing translation for 'onlineSavingsMsg'
Learn More

Tri(2-furyl)phosphine 98.0+%, TCI America™

Catalog No. T16435G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16435G 5 g
T16431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16435G Supplier TCI America Supplier No. T16435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5518-52-5
Molecular Formula C12H9O3P
Molecular Weight (g/mol) 232.18
MDL Number MFCD00151857
InChI Key DLQYXUGCCKQSRJ-UHFFFAOYSA-N
Synonym tri 2-furyl phosphine, tri furan-2-yl phosphine, tri-2-furyl phosphine, tris furan-2-yl phosphane, tri-2-furylphosphine, tris 2-furyl phosphine, tri furan-2-yl phosphane, trifurylphosphine, tri fur-2-yl phosphane, tris 2-furanyl phosphine
PubChem CID 521585
IUPAC Name tris(furan-2-yl)phosphane
SMILES O1C=CC=C1P(C1=CC=CO1)C1=CC=CO1

Specifications

Melting Point 63°C
Boiling Point 136°C
Quantity 5 g
Formula Weight 232.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tri(2-furyl)phosphine