accessibility menu, dialog, popup

UserName

H3CHOHO(S)CH3(R)HOOOCH2(E)

CAS RN 5508-58-7

IUPAC Name: (3E)-3-{2-[(1R,8aS)-6-hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylidene-decahydronaphthalen-1-yl]ethylidene}-4-hydroxyoxolan-2-one
Synonyms: andrographolide 穿心莲内酯
Molecular Weight (g/mol): 350.46
Molecular Formula: C20H30O5
InChi Key: BOJKULTULYSRAS-ZJFCSBQFSA-N
SMILES: CC1(CO)C(O)CC[C@@]2(C)[C@H](C\C=C3/C(O)COC3=O)C(=C)CCC12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results