accessibility menu, dialog, popup

UserName

CAS RN 55-56-1

IUPAC Name: N'-(6-{[amino({amino[(4-chlorophenyl)amino]methylidene}amino)methylidene]amino}hexyl)-1-({amino[(4-chlorophenyl)amino]methylidene}amino)methanimidamide
Synonyms: chlorhexidine
Molecular Weight (g/mol): 505.45
Molecular Formula: C22H30Cl2N10
InChi Key: GHXZTYHSJHQHIJ-UHFFFAOYSA-N
SMILES: NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)