missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine, 98%, Thermo Scientific Chemicals

Catalog No. AC461020050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461020050 5 g
AC461020010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461020050 Supplier Thermo Scientific Chemicals Supplier No. 461020050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55-56-1
Molecular Formula C22H30Cl2N10
Molecular Weight (g/mol) 505.45
InChI Key GHXZTYHSJHQHIJ-UHFFFAOYSA-N
Synonym chlorhexidine, chlorhexidinum, chlorhexidin, nolvasan, rotersept, tubulicid, fimeil, hexadol, soretol, cloresidina
PubChem CID 9552079
ChEBI CHEBI:3614
IUPAC Name (1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine
SMILES C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl

Specifications

Melting Point 134.0°C to 136.0°C
Quantity 5 g
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 505.45
Percent Purity 98%
Chemical Name or Material Chlorhexidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes serious eye damage. Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 200-238-7

RUO – Research Use Only