missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019614
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019614 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019614 Supplier Merck Emd Millipore Supplier No. PHR14211G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Chlorhexidine, the most popular antiseptic of biguanides, acts as a potential antimicrobial agent against several gram-positive and a few gram-negative bacteria. It is prescribed as a medicine for the prevention and treatment of the redness, swelling, and bleeding gums associated with gingivitis in humans. It may also be used as a general-purpose disinfectant for cleansing wounds, skin, instrument, and equipment in veterinary applications. Other applications of chlorhexidine include its use as the component of shampoos, ointments, skin and wound cleansers etc.

Specifications

CAS 55-56-1
Physical Form Neat
Linear Formula [-(CH2)3 NHC(=NH)NHC(=NH)NHC6H4Cl]2
MDL Number MFCD00009673
Melting Point 134°C to 136°C
Synonym Chlorhexidine; 1,1 '-Hexamethylenebis[5-(4-chlorophenyl)biguanide]
Quantity 1 g
Formula Weight 505.45
Grade Certified Reference Material