accessibility menu, dialog, popup

UserName

CH3BrCl

CAS RN 54932-72-8

IUPAC Name: 4-bromo-1-chloro-2-methylbenzene
Synonyms: 4-bromo-1-chloro-2-methylbenzene
Molecular Weight (g/mol): 205.48
Molecular Formula: C7H6BrCl
InChi Key: OZFQMHJKAODEON-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=CC(Br)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)