accessibility menu, dialog, popup

UserName

CAS RN 548-35-6

IUPAC Name: heptacyclo[18.7.0.0²,¹⁰.0³,⁸.0¹¹,¹⁹.0¹²,¹⁷.0²¹,²⁶]heptacosa-1(20),2(10),3,5,7,11(19),12,14,16,21,23,25-dodecaene
Synonyms: heptacyclo[18.7.0.0²,¹⁰.0³,⁸.0¹¹,¹⁹.0¹²,¹⁷.0²¹,²⁶]heptacosa-1(20),2(10),3,5,7,11(19),12,14,16,21,23,25-dodecaene
Molecular Weight (g/mol): 342.44
Molecular Formula: C27H18
InChi Key: YGPLLMPPZRUGTJ-UHFFFAOYSA-N
SMILES: C1C2=CC=CC=C2C2=C1C1=C(CC3=CC=CC=C13)C1=C2CC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)