missing translation for 'onlineSavingsMsg'
Learn More

Truxene 98.0+%, TCI America™

Catalog No. T27521G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27521G 1 g
T2752100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27521G Supplier TCI America Supplier No. T27521G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 548-35-6
Molecular Formula C27H18
Molecular Weight (g/mol) 342.441
MDL Number MFCD00021180
InChI Key YGPLLMPPZRUGTJ-UHFFFAOYSA-N
PubChem CID 68355
SMILES C1C2=CC=CC=C2C3=C1C4=C(CC5=CC=CC=C54)C6=C3CC7=CC=CC=C76

Specifications

Melting Point 370°C
Color Yellow
Quantity 1 g
Synonym truxene, truxen, .alpha.-truxene, 5h-tribenzo a,f,k trindene, 10,15-dihydro, benzene, tribenzylene, 10,15-dihydro-5h-tribenzo a,f,k triindene, 10,15-dihydro-5h-diindeno 1,2-a:1',2'-c fluorene, 5h-tribenzo a,f,k trindene,10,15-dihydro, tribenzylenebenzene
Formula Weight 342.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Truxene