accessibility menu, dialog, popup

UserName

CAS RN 54716-02-8

IUPAC Name: ethyl (2E)-3-(pyrrolidin-1-yl)but-2-enoate
Synonyms: ethyl (2E)-3-(pyrrolidin-1-yl)but-2-enoate
Molecular Weight (g/mol): 183.25
Molecular Formula: C10H17NO2
InChi Key: MSOQKPXSIHLODG-CMDGGOBGSA-N
SMILES: CCOC(=O)\C=C(/C)N1CCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)