accessibility menu, dialog, popup

UserName

CAS RN 5470-17-7

IUPAC Name: 3-bromo-2-chloro-5-nitropyridine
Synonyms: 3-bromo-2-chloro-5-nitropyridine
Molecular Weight (g/mol): 237.44
Molecular Formula: C5H2BrClN2O2
InChi Key: PTTQIUHVDDBART-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CN=C(Cl)C(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results