accessibility menu, dialog, popup

UserName

CAS RN 5467-74-3

IUPAC Name: (4-bromophenyl)boronic acid
Synonyms: 4-bromophenylboric acid
Molecular Weight (g/mol): 200.83
Molecular Formula: C6H6BBrO2
InChi Key: QBLFZIBJXUQVRF-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(Br)C=C1
Quantity 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)