missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B18581G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B18581G Supplier TCI America Supplier No. B18581G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5467-74-3
Molecular Formula C6H6BBrO2
Molecular Weight (g/mol) 200.83
MDL Number MFCD00002104
InChI Key QBLFZIBJXUQVRF-UHFFFAOYSA-N
Synonym 4-bromophenyl boronic acid, 4-bromobenzeneboronic acid, 4-bromophenylboric acid, p-bromophenylboronic acid, p-bromophenylboric acid, p-bromobenzeneboronic acid, boronic acid, 4-bromophenyl, benzeneboronic acid, p-bromo, p-bromophenyl boronic acid, 4-bromo phenyl boronic acid
PubChem CID 79599
IUPAC Name (4-bromophenyl)boronic acid
SMILES OB(O)C1=CC=C(Br)C=C1

Specifications

Melting Point 292°C
Quantity 1 g
Formula Weight 200.83
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophenylboronic Acid (contains varying amounts of Anhydride)
RTECSNumber CY8650000
TSCA No