accessibility menu, dialog, popup

UserName

CH3CH3CH3H3CNOONOO

CAS RN 5465-13-4

IUPAC Name: 1,2,4,5-tetramethyl-3,6-dinitrobenzene
Synonyms: 1,2,4,5-tetramethyl-3,6-dinitrobenzene
Molecular Weight (g/mol): 224.22
Molecular Formula: C10H12N2O4
InChi Key: AEPQXGFMAZTUEA-UHFFFAOYSA-N
SMILES: CC1=C(C)C(=C(C)C(C)=C1[N+]([O-])=O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)