missing translation for 'onlineSavingsMsg'
Learn More

Dinitrodurene 97.0+%, TCI America™

Numéro de catalogue. D166125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D166125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. D166125G Fournisseur TCI America Code fournisseur D166125G
Il en reste null

Identifiants chimiques

CAS 5465-13-4
Molecular Formula C10H12N2O4
Molecular Weight (g/mol) 224.216
MDL Number MFCD00007164
InChI Key AEPQXGFMAZTUEA-UHFFFAOYSA-N
Synonym 1,2,4,5-tetramethyl-3,6-dinitro-benzene, dinitrotrodurene, 3,2,4,5-tetramethylbenzene, 2,3,5,6-tetramethyl-p-dinitrobenzene, 1,4,5-tetramethyl-3,6-dinitrobenzene, benzene,2,4,5-tetramethyl-3,6-dinitro, 1,4-dinitro-2,3,5,6-tetramethylbenzene 97+%
PubChem CID 79593
IUPAC Name 1,2,4,5-tetramethyl-3,6-dinitrobenzene
SMILES CC1=C(C(=C(C(=C1[N+](=O)[O-])C)C)[N+](=O)[O-])C

Spécifications

Melting Point 210°C
Color Yellow
Quantity 25 g
Formula Weight 224.22
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dinitrodurene