missing translation for 'onlineSavingsMsg'
Learn More

Dinitrodurene, 97%, Thermo Scientific™

Catalog No. p-7077146
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC117000010 1g
AC117000050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC117000010 Supplier Thermo Scientific Chemicals Supplier No. 117000010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5465-13-4
Molecular Formula C10H12N2O4
Molecular Weight (g/mol) 224.22
InChI Key AEPQXGFMAZTUEA-UHFFFAOYSA-N
Synonym 1,2,4,5-tetramethyl-3,6-dinitro-benzene, dinitrotrodurene, 3,2,4,5-tetramethylbenzene, 2,3,5,6-tetramethyl-p-dinitrobenzene, 1,4,5-tetramethyl-3,6-dinitrobenzene, benzene,2,4,5-tetramethyl-3,6-dinitro, 1,4-dinitro-2,3,5,6-tetramethylbenzene 97+%
PubChem CID 79593
IUPAC Name 1,2,4,5-tetramethyl-3,6-dinitrobenzene
SMILES CC1=C(C(=C(C(=C1[N+](=O)[O-])C)C)[N+](=O)[O-])C

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)4C6(NO2)2
Quantity 1g
Beilstein 05,433
Formula Weight 224.22
Percent Purity 97%
Chemical Name or Material Dinitrodurene
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Store in a
EINECSNumber 226-766-8

RUO – Research Use Only