accessibility menu, dialog, popup

UserName

CH3NCH3CH3HOCH3NCH3CH3HOCH3NCH3CH3HOOHOOOOOO

CAS RN 546-63-4

IUPAC Name: tris((2-hydroxyethyl)trimethylazanium) 2-hydroxypropane-1,2,3-tricarboxylate
Synonyms: tris(choline) citrate
Molecular Weight (g/mol): 501.62
Molecular Formula: C21H47N3O10
InChi Key: RPERJPYDELTDMR-UHFFFAOYSA-K
SMILES: C[N+](C)(C)CCO.C[N+](C)(C)CCO.C[N+](C)(C)CCO.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results