accessibility menu, dialog, popup

UserName

CAS RN 544-85-4

IUPAC Name: dotriacontane
Synonyms: dotriacontane bicetyl
Molecular Weight (g/mol): 450.88
Molecular Formula: C32H66
InChi Key: QHMGJGNTMQDRQA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Boiling Point 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)