accessibility menu, dialog, popup

UserName

H3CCl

CAS RN 544-10-5

IUPAC Name: 1-chlorohexane
Synonyms: 1-chlorohexane
Molecular Weight (g/mol): 120.62
Molecular Formula: C6H13Cl
InChi Key: MLRVZFYXUZQSRU-UHFFFAOYSA-N
SMILES: CCCCCCCl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)