accessibility menu, dialog, popup

UserName

CAS RN 543-87-3

IUPAC Name: 3-methylbutyl nitrate
Synonyms: isopentyl nitrate
Molecular Weight (g/mol): 133.15
Molecular Formula: C5H11NO3
InChi Key: NTHGIYFSMNNHSC-UHFFFAOYSA-N
SMILES: CC(C)CCO[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results