accessibility menu, dialog, popup

UserName

CAS RN 54060-30-9

IUPAC Name: 3-ethynylaniline
Synonyms: 3-ethynylaniline
Molecular Weight (g/mol): 117.15
Molecular Formula: C8H7N
InChi Key: NNKQLUVBPJEUOR-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)C#C
Boiling Point 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (3)