accessibility menu, dialog, popup

UserName

CAS RN 540-54-5

IUPAC Name: 1-chloropropane
Synonyms: 1-chloropropane N-propyl chloride
Molecular Weight (g/mol): 78.54
Molecular Formula: C3H7Cl
InChi Key: SNMVRZFUUCLYTO-UHFFFAOYSA-N
SMILES: CCCCl
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)