accessibility menu, dialog, popup

UserName

CAS RN 54-25-1

IUPAC Name: 2-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2,3,4,5-tetrahydro-1,2,4-triazine-3,5-dione
Synonyms: 6-azauridine
Molecular Weight (g/mol): 245.19
Molecular Formula: C8H11N3O6
InChi Key: WYXSYVWAUAUWLD-UHFFFAOYNA-N
SMILES: OCC1OC(C(O)C1O)N1N=CC(=O)NC1=O
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)