CAS RN 5398-27-6
CAS RN 5398-27-6
IUPAC Name:
2,6-diiodo-4-nitroaniline
Synonyms:
2,6-diiodo-4-nitroaniline
Molecular Weight (g/mol):
389.92
Molecular Formula:
C6H4I2N2O2
InChi Key:
YPVYMWQYENWFAT-UHFFFAOYSA-N
SMILES:
NC1=C(I)C=C(C=C1I)[N+]([O-])=O
1
–
1
of
1
results
1
2,6-Diiodo-4-nitroaniline 98.0+%, TCI America™
CAS: 5398-27-6 Molecular Formula: C6H4I2N2O2 Molecular Weight (g/mol): 389.919 MDL Number: MFCD00007683 InChI Key: YPVYMWQYENWFAT-UHFFFAOYSA-N PubChem CID: 79386 IUPAC Name: 2,6-diiodo-4-nitroaniline SMILES: C1=C(C=C(C(=C1I)N)I)[N+](=O)[O-]