missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Diiodo-4-nitroaniline 98.0+%, TCI America™
$277.75 - $1112.84
Chemical Identifiers
| CAS | 5398-27-6 |
|---|---|
| Molecular Formula | C6H4I2N2O2 |
| Molecular Weight (g/mol) | 389.919 |
| MDL Number | MFCD00007683 |
| InChI Key | YPVYMWQYENWFAT-UHFFFAOYSA-N |
| PubChem CID | 79386 |
| IUPAC Name | 2,6-diiodo-4-nitroaniline |
| SMILES | C1=C(C=C(C(=C1I)N)I)[N+](=O)[O-] |
Chemical Identifiers
| 5398-27-6 | |
| 389.919 | |
| YPVYMWQYENWFAT-UHFFFAOYSA-N | |
| 2,6-diiodo-4-nitroaniline |
| C6H4I2N2O2 | |
| MFCD00007683 | |
| 79386 | |
| C1=C(C=C(C(=C1I)N)I)[N+](=O)[O-] |
Specifications
| 5398-27-6 | |
| C6H4I2N2O2 | |
| 5 g | |
| C1=C(C=C(C(=C1I)N)I)[N+](=O)[O-] | |
| 389.919 | |
| 389.92 | |
| Crystalline Powder |
| 255°C | |
| MFCD00007683 | |
| YPVYMWQYENWFAT-UHFFFAOYSA-N | |
| 2,6-diiodo-4-nitroaniline | |
| 79386 | |
| ≥98.0% (GC) | |
| 2,6-Diiodo-4-nitroaniline |
Safety and Handling
TSCA : No