accessibility menu, dialog, popup

UserName

H3CNNNNSONHNNSH3COO

CAS RN 5379-16-8

IUPAC Name: N-[4-(4-methanesulfonylpiperazin-1-yl)phenyl]-2-{[1-(2-methylphenyl)-1H-1,2,3,4-tetrazol-5-yl]sulfanyl}acetamide
Synonyms: N-[4-(4-methanesulfonylpiperazin-1-yl)phenyl]-2-{[1-(2-methylphenyl)-1,2,3,4-tetrazol-5-yl]sulfanyl}acetamide
Molecular Weight (g/mol): 487.6
Molecular Formula: C21H25N7O3S2
InChi Key: FDAKJDJSCMDVPB-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1N1N=NN=C1SCC(=O)NC1=CC=C(C=C1)N1CCN(CC1)S(C)(=O)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)