accessibility menu, dialog, popup

UserName

CAS RN 537-40-6

H3COOOOCH3OOH3C(Z)(Z)(Z)(Z)(Z)(Z)

CAS RN 537-40-6

IUPAC Name: 1,3-bis[(9Z,12Z)-octadeca-9,12-dienoyloxy]propan-2-yl (9Z,12Z)-octadeca-9,12-dienoate
Synonyms: triglyceride
Molecular Weight (g/mol): 879.41
Molecular Formula: C57H98O6
InChi Key: HBOQXIRUPVQLKX-BBWANDEASA-N
SMILES: CCCCC\C=C/C\C=C/CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/C\C=C/CCCCC)OC(=O)CCCCCCC\C=C/C\C=C/CCCCC
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results