missing translation for 'onlineSavingsMsg'
Learn More

Glyceryl trilinoleate, Analytical Standard, ≥97.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014518
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014518 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014518 Supplier Merck Emd Millipore Supplier No. 96373100MG

Specifications

CAS 537-40-6
Density 0.925 g/mL (at 20°C)
Physical Form Neat
Linear Formula C57H98O6
MDL Number MFCD00042909
Synonym 1,2,3-Tri-(cis,cis-9,12-octadecadienoyl)glycerol; 1,2,3-Trilinoleoylglycerol; Glycerol trilinoleate; Trilinolein
Quantity 100 mg
Formula Weight 879.38
Percent Purity ≥97.5% (GC)
Grade Analytical Standard