accessibility menu, dialog, popup

UserName

CAS RN 5351-23-5

IUPAC Name: 4-hydroxybenzohydrazide
Synonyms: benzoylhydrazine, P-hydroxy
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: ZMZGIVVRBMFZSG-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (5)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)