accessibility menu, dialog, popup

UserName

CAS RN 533-18-6

IUPAC Name: 2-methylphenyl acetate
Synonyms: O-cresyl acetate
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: AMZORBZSQRUXNC-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC=C1C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results