accessibility menu, dialog, popup

UserName

CAS RN 53218-26-1

IUPAC Name: 6-bromo-1,3-benzothiazole
Synonyms: 6-bromo-1,3-benzothiazole
Molecular Weight (g/mol): 214.08
Molecular Formula: C7H4BrNS
InChi Key: YJOUISWKEOXIMC-UHFFFAOYSA-N
SMILES: BrC1=CC=C2N=CSC2=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)