accessibility menu, dialog, popup

UserName

ONCS

CAS RN 532-55-8

IUPAC Name: benzoyl isothiocyanate
Synonyms: benzoyl isothiocyanate
Molecular Weight (g/mol): 163.19
Molecular Formula: C8H5NOS
InChi Key: CPEKAXYCDKETEN-UHFFFAOYSA-N
SMILES: O=C(N=C=S)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (4)