accessibility menu, dialog, popup

UserName

PH3CH3CH3CH3CCH3CH3H3CCH3CH3PCH3CH3CH3CH3H3CCH3CH3H3CH3CPd

CAS RN 53199-31-8

IUPAC Name: bis(tri-tert-butylphosphane) palladium
Synonyms: bis(tri-tert-butylphosphane) palladium
Molecular Weight (g/mol): 511.06
Molecular Formula: C24H54P2Pd
InChi Key: MXQOYLRVSVOCQT-UHFFFAOYSA-N
SMILES: [Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C
Physical Form 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)