accessibility menu, dialog, popup

UserName

H3COOONH2

CAS RN 53171-39-4

IUPAC Name: methyl 3-carbamoylpropanoate
Synonyms: methyl 3-carbamoylpropanoate
Molecular Weight (g/mol): 131.13
Molecular Formula: C5H9NO3
InChi Key: HURZMSZDVGMYKJ-UHFFFAOYSA-N
SMILES: COC(=O)CCC(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results