accessibility menu, dialog, popup

UserName

CH3OOOCH3NOO

CAS RN 5307-17-5

IUPAC Name: methyl 3-methoxy-2-nitrobenzoate
Synonyms: methyl 3-methoxy-2-nitrobenzoate
Molecular Weight (g/mol): 211.17
Molecular Formula: C9H9NO5
InChi Key: FDQQRLPHAAICCR-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(OC)=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)