accessibility menu, dialog, popup

UserName

CAS RN 52933-01-4

IUPAC Name: butoxyphosphonic acid; dibutoxyphosphinic acid
Synonyms: butoxyphosphonic acid; di-N-butyl phosphate
Molecular Weight (g/mol): 364.31
Molecular Formula: C12H30O8P2
InChi Key: ORMYKVYVDRWAKI-UHFFFAOYSA-N
SMILES: CCCCOP(O)(O)=O.CCCCOP(O)(=O)OCCCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results