accessibility menu, dialog, popup

UserName

CAS RN 52768-23-7

IUPAC Name: 6,7-dihydroxy-1,2,3,4-tetrahydroisoquinolin-2-ium
Synonyms: 6,7-dihydroxy-1,2,3,4-tetrahydroisoquinolin-2-ium
Molecular Weight (g/mol): 166.2
Molecular Formula: C9H12NO2
InChi Key: MBFUSGLXKQWVDW-UHFFFAOYSA-O
SMILES: OC1=CC2=C(C[NH2+]CC2)C=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)