accessibility menu, dialog, popup

UserName

CAS RN 5274-70-4

IUPAC Name: 2-hydroxy-3-nitrobenzaldehyde
Synonyms: 2-hydroxy-3-nitrobenzaldehyde
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: NUGOTBXFVWXVTE-UHFFFAOYSA-N
SMILES: OC1=C(C=O)C=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)