accessibility menu, dialog, popup

UserName

H3CN

CAS RN 52709-84-9

IUPAC Name: 4'-octyl-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-octyl-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 291.44
Molecular Formula: C21H25N
InChi Key: CSQPODPWWMOTIY-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)