missing translation for 'onlineSavingsMsg'
Learn More

4'-n-Octylbiphenyl-4-carbonitrile, 99%, Thermo Scientific Chemicals

Catalog No. AAH2631803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2631803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2631803 Supplier Thermo Scientific Chemicals Supplier No. H2631803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52709-84-9 | Molecular Formula: C21H25N | Molecular Weight: 291.44 g/mol

The thermobarometry of thermotropic liquid crystals is feasible by the metal tube Berthelot method, and its extension in the negative-pressure range will be interesting for the study of organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The thermobarometry of thermotropic liquid crystals is feasible by the metal tube Berthelot method, and its extension in the negative-pressure range will be interesting for the study of organic compounds.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.

Chemical Identifiers

CAS 52709-84-9
Molecular Formula C21H25N
Molecular Weight (g/mol) 291.44
MDL Number MFCD00075146
InChI Key CSQPODPWWMOTIY-UHFFFAOYSA-N
Synonym 4-cyano-4'-octylbiphenyl, 4'-octyl-1,1'-biphenyl-4-carbonitrile, 4-4-octylphenyl benzonitrile, 1,1'-biphenyl-4-carbonitrile, 4'-octyl, 4-octylcyanodiphenyl, k 24 liquid crystal, cb 8, 4'-octyl-4-biphenylcarbonitrile, 4'-octyl 1,1'-biphenyl-4-carbonitrile, 4-octyl-4'-cyanobiphenyl
PubChem CID 104289
IUPAC Name 4-(4-octylphenyl)benzonitrile
SMILES CCCCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N

Specifications

Melting Point 20°C to 22°C
Flash Point >110°C (230°F)
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 291.43
Percent Purity 99%
Chemical Name or Material 4'-n-Octylbiphenyl-4-carbonitrile
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 258-120-6
RTECSNumber DV1764400
TSCA Yes

RUO – Research Use Only