missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-n-octylbiphenyl 98.0+%, TCI America™

Catalog No. C31565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31565G 5 g
C315625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31565G Supplier TCI America Supplier No. C31565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52709-84-9
Molecular Formula C21H25N
Molecular Weight (g/mol) 291.44
MDL Number MFCD00075146
InChI Key CSQPODPWWMOTIY-UHFFFAOYSA-N
Synonym 4-cyano-4'-octylbiphenyl, 4'-octyl-1,1'-biphenyl-4-carbonitrile, 4-4-octylphenyl benzonitrile, 1,1'-biphenyl-4-carbonitrile, 4'-octyl, 4-octylcyanodiphenyl, k 24 liquid crystal, cb 8, 4'-octyl-4-biphenylcarbonitrile, 4'-octyl 1,1'-biphenyl-4-carbonitrile, 4-octyl-4'-cyanobiphenyl
PubChem CID 104289
IUPAC Name 4'-octyl-[1,1'-biphenyl]-4-carbonitrile
SMILES CCCCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N

Specifications

Color White
Boiling Point 218°C
Quantity 5 g
Formula Weight 291.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-n-octylbiphenyl
RTECSNumber DV1764400
TSCA Yes