accessibility menu, dialog, popup

UserName

ClOO

CAS RN 527-69-5

IUPAC Name: furan-2-carbonyl chloride
Synonyms: furoyl chloride
Molecular Weight (g/mol): 130.53
Molecular Formula: C5H3ClO2
InChi Key: OFTKFKYVSBNYEC-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CO1
Boiling Point 
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)