missing translation for 'onlineSavingsMsg'
Learn More

2-Furoyl Chloride 98.0+%, TCI America™

Catalog No. F0233500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0233500G 500 g
F023325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0233500G Supplier TCI America Supplier No. F0233500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 527-69-5
Molecular Formula C5H3ClO2
Molecular Weight (g/mol) 130.527
MDL Number MFCD00003228
InChI Key OFTKFKYVSBNYEC-UHFFFAOYSA-N
Synonym 2-furoyl chloride, furoyl chloride, 2-furancarbonyl chloride, furancarbonyl chloride, 2-furoylchloride, unii-2us0dxi75s, 2us0dxi75s, 2-furancarboxylic acid chloride, furan-2-carboxylic acid chloride, furanoyl chloride
PubChem CID 68242
IUPAC Name furan-2-carbonyl chloride
SMILES C1=COC(=C1)C(=O)Cl

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 174°C
Quantity 500 g
UN Number 3265
Formula Weight 130.53
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Furoyl Chloride
RTECSNumber LT9925000
TSCA Yes