missing translation for 'onlineSavingsMsg'
Learn More

2-Furoyl chloride, 98+%

Catalog No. AAL0696030
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0696030 250 g
AAL0696018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0696030 Supplier Thermo Scientific Chemicals Supplier No. L0696030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 527-69-5 | C5H3ClO2 | 130.527 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 527-69-5
Molecular Formula C5H3ClO2
Molecular Weight (g/mol) 130.527
MDL Number MFCD00003228
InChI Key OFTKFKYVSBNYEC-UHFFFAOYSA-N
Synonym 2-furoyl chloride, furoyl chloride, 2-furancarbonyl chloride, furancarbonyl chloride, 2-furoylchloride, unii-2us0dxi75s, 2us0dxi75s, 2-furancarboxylic acid chloride, furan-2-carboxylic acid chloride, furanoyl chloride
PubChem CID 68242
IUPAC Name furan-2-carbonyl chloride
SMILES C1=COC(=C1)C(=O)Cl

Specifications

Melting Point -2°C to +1°C
Density 1.325
Boiling Point 173°C to 174°C
Flash Point 85°C (185°F)
Refractive Index 1.531
Quantity 250 g
UN Number UN3265
Beilstein 110144
Sensitivity Moisture sensitive
Merck Index 14,4310
Formula Weight 130.53
Percent Purity ≥98%
Chemical Name or Material 2-Furoyl chloride
Show More Show Less
Hazard Category H227-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 208-422-9
RTECSNumber LT9925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only