accessibility menu, dialog, popup

UserName

H3CONIOH3CN

CAS RN 52522-99-3

IUPAC Name: 5-iodo-2,4-dimethoxypyrimidine
Synonyms: 5-iodo-2,4-dimethoxypyrimidine
Molecular Weight (g/mol): 266.04
Molecular Formula: C6H7IN2O2
InChi Key: KNTMOGOYRYYUIE-UHFFFAOYSA-N
SMILES: COC1=NC=C(I)C(OC)=N1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)