accessibility menu, dialog, popup

UserName

BrOPO

CAS RN 52509-14-5

IUPAC Name: [(1,3-dioxolan-2-yl)methyl]triphenylphosphanium bromide
Synonyms: (1,3-dioxolan-2-ylmethyl)triphenylphosphanium bromide
Molecular Weight (g/mol): 429.29
Molecular Formula: C22H22BrO2P
InChi Key: FRHRVQQUICVJDG-UHFFFAOYSA-M
SMILES: [Br-].C(C1OCCO1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)