missing translation for 'onlineSavingsMsg'
Learn More

(1,3-Dioxolan-2-yl)methyltriphenylphosphonium Bromide 97.0+%, TCI America™

Catalog No. D21645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21645G 5 g
D216425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21645G Supplier TCI America Supplier No. D21645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52509-14-5
Molecular Formula C22H22BrO2P
Molecular Weight (g/mol) 429.29
MDL Number MFCD00011966
InChI Key FRHRVQQUICVJDG-UHFFFAOYSA-M
Synonym 1,3-dioxolan-2-ylmethyl triphenylphosphonium bromide, 1,3-dioxolan-2-yl methyl triphenylphosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenylphosphanium bromide, phosphonium, 1,3-dioxolan-2-ylmethyl triphenyl-, bromide, 1,3-dioxolan-2-yl methyltriphenylphosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenyl phosphanium bromide, 1,3-dioxolan-2-ylmethyl triphenyl phosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenylphosphine, bromide, acmc-209kzy
PubChem CID 2724194
IUPAC Name [(1,3-dioxolan-2-yl)methyl]triphenylphosphanium bromide
SMILES [Br-].C(C1OCCO1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 191°C
Quantity 5 g
Formula Weight 429.29
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1,3-Dioxolan-2-yl)methyltriphenylphosphonium Bromide
EINECSNumber (5)-6734
TSCA No